Human Recombinant Endocrine Gland VEGF
Supplier: Rockland Immunochemical
009-001-U74-0100 009-001-U74-0020 009-001-U74-0005
10802-606EA 4839 USD
10802-606 10802-608 10802-610 10802-612
Human Recombinant Endocrine Gland VEGF
Proteins and Peptides
Recombinant Human Endocrine Gland VEGF control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Endocrine Gland-derived Vascular Endothelial Growth Factor (EG-VEGF) is an angiogenic growth factor specifically expressed in the ovaries, testis, adrenal and placental tissues. The identification of tissue-selective angiogenic factors raises the possibility that other secreted molecules in this class exist. EG-VEGF expression correlates with vascularity in polycystic ovary syndrome, a leading cause of infertility. Recombinant human EG-VEGF is a non-glycosylated protein, containing 86 amino acids, with a molecular weight of 9.6 kDa.
Endocrine Gland Vascular Endothelial Growth Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Endocrine Gland Vascular Endothelial Growth Factor in immunological assays.