Anti-KLHL25 Chicken Polyclonal Antibody

Supplier: Rockland Immunochemical
200-901-BA4
10802-604EA 635.22 USD
10802-604
Anti-KLHL25 Chicken Polyclonal Antibody
Antibodies
The ectoderm-neural cortex-2 (ENC-2) protein a member of the BTB/PDZ Kelch protein family displaying significant homology to ENC-1, an actin-binding protein involved in neural fate specification during embryogenesis and adipocyte maturation. The exact function of ENC-2 has yet to be determined, but it has been suggested to be involved in the differentiation of neuronal precursor cells into neurons and astrocytes.

Recommended Dilutions: ELISA: 1:10,000; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: KLHL25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Chicken
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR