Anti-C2ORF40 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-AY9
10802-568EA 635.22 USD
10802-568
Anti-C2ORF40 Rabbit Polyclonal Antibody
Antibodies
The esophageal cancer-related gene 4 (ECRG4), also known as Augurin, was identified through differential display screening between esophageal cancer and normal tissue. Hypermethylation of CpG sites in the ECRG4 promoter correlate with ECRG4 downregulation in numerous esophageal squamous cell carcinoma tissues.. In transfected cells, overexpression of ECRG4 induced G1 arrest, dephosphorylation of Rb and decreased expression of cyclins D1 and D3. Increased expression of ECRG4 in aged mouse brain is observed compared with young mouse brain. The increased ECRG4 level was accompanied by elevated senescence-associated acidic beta-galactosidase activity, suggesting that ECRG4 is involved in cellular differentiation and senescence.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: C2ORF40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR