DONKEY SERUM (Non Sterile), Rockland Immunochemicals
Supplier: Rockland Immunochemical
D220-00-0050 D220-00-0100
10802-530EA 366.72 USD
10802-530 10802-532
DONKEY SERUM (Non Sterile), Rockland Immunochemicals
Sera (Serum)
Non-Sterile Serum
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Donkey serum is used as a component of bioassays, immunoassays or enzyme assays. Donkey serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Donkey serum is also available as a sterile preparation for use as a cell culture supplement.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal