Anti-Choline Oxidase Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
200-106-094S
10802-382EA 151.49 USD
10802-382
Anti-Choline Oxidase Goat Polyclonal Antibody (Biotin)
Antibodies
Choline Oxidase is a flavoprotein responsible for the catalyzing the conversion of choline to betaine aldehyde in animals and bacteria. Anti-Choline Oxidase antibody is ideal for researchers interested in Metabolism and Signal Transduction research.

Recommended Dilutions: ELISA: 1:1,000 - 1:2,500; Immunoprecipitation: 1:100; Western Blot: 1:500 - 1:1,000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: Choline Oxidase
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Alcaligenes Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR