CHO/K1 Whole Cell Lysate, Rockland Immunochemical
Supplier: Rockland Immunochemical
W07-000-359
10802-374EA
177.61
USD
10802-374
CHO/K1 Whole Cell Lysate, Rockland Immunochemical
Protein Extraction
Protein Extraction Reagents
Ready-to-use whole cell lysates are derived from cell lines or tissues using highly refined extraction protocols to ensure exceptionally high quality, protein integrity and lot-to-lot reproducibility.
- Synonyms: CHO/K1 Whole Cell Lysate, CHO K1 WCL, CHO-K1 lysate, CHOK1 lysate
- Stabilizer: 10% (v/v) Glycerol
- Origin: Chinese Hamster
- Cell Line: Chinese Hamster Ovary (CHO)
1X SDS-PAGE Sample Buffer (62.5 mM Tris HCl, 2% SDS, 10% Glycerol and 0.005% bromophenol blue, pH 6.8) is used.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...