Bovine Serum Albumin (BSA)
Supplier: Rockland Immunochemical
001-0133-0010
10802-172EA 120.15 USD
10802-172
Bovine Serum Albumin (BSA)
Proteins and Peptides
Bovine Serum Albumin (BSA)
  • protein standard
Bovine Serum Albumin (BSA) is used for various biochemical applications including ELISA (Enzyme-Linked Immunosorbent Assay), high content screening assays, western blotting, FACS Buffer and immunohistochemistry. Also used as a nutrient in cell and microbial culture. In restriction digests, BSA is used to stabilize some enzymes during digestion of DNA and to prevent adhesion of the enzyme to reaction tubes and other vessels. Bovine Serum Albumin can also be used to determine the quantity of other proteins, by comparing an unknown quantity of protein to known amounts of BSA.
Bovine Serum Albumin (BSA) is suitable for use in protease sensitive assays such as RIA, EIA and nucleic acid hybridization, use as a stabilizing agent for proteins and enzymes including dilute solutions of antibody, use as a blocking agent to reduce non-specific binding.