BBS Fish Gel Concentrate 10X , Rockland Immunochemicals
Supplier: Rockland Immunochemical
MB-065-0100 MB-065-1000
10802-022EA 120.15 USD
10802-022 10802-024
BBS Fish Gel Concentrate 10X , Rockland Immunochemicals
Assays ELISAs
Borate Buffered Saline Fish Gel Concentrate 10X
  • 10X concentrated stock solution
  • 1.0M boric acid, 1.5M sodium chloride and Fish Gelatin at pH 8.0
  • minimal non-specific interactions
  • minimize background
Highly sensitive ELISA assays require minimal non-specific interactions.  Non-specific binding results in elevated background levels and a decrease in signal-to-noise ratios.  Blocking reagents act to minimize non-specific interactions of secondary reactants with each other and with the primary solid phase binding sites. Rockland offers several options for antibody diluents in multiple buffer configurations that effectively block non-specific interactions and minimize background. Multiple formulations offer greater flexibility in experimental design.
This product is a 10X concentrated stock solution.  Prepare a 1X working solution by diluting 1 part 10X concentrate with 9 parts distilled-deionized water or equivalent. 10X BBS Fish Gel Concentrate consists of 1.0M boric acid, 1.5M sodium chloride and Fish Gelatin at pH 8.0.  A proprietary combination of stabilizers and preservatives are used that are azide and mercury free.