Anti-BCAP29 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-Z28
10802-000EA 635.22 USD
10802-000
Anti-BCAP29 Rabbit Polyclonal Antibody
Antibodies
Bap29 and the related protein Bap31 are endoplasmic reticulum (ER) and ER-vesicle membrane proteins and members of the B-cell receptor-associated protein family. These two proteins are highly homologous and can form homo- and heterodimers. Both Bap29 and Bap31 interact with membrane-bound immunoglobulins (mIgs), such as IgM and IgD, which with Ig-alpha/Ig-beta heterodimers form B cell antigen receptors. Binding of the Bap29/Bap31 heterodimer correlates with the ER retention of non-Ig-alpha/Ig-beta bound mIg complexes, suggesting that Bap29 and Bap31 may act as chaperones transmembrane regions of various proteins. Bap29 possesses multiple isoforms.

Recommended Dilutions: ELISA: 1:5000-1:20,000; Immunohistochemsitry: 10 µg/mL; Immunofluorescence Microscopy: 20 µg/mL; Western Blot: 0.5 - 2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: BCAP29
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR