Anti-GPD1 Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
200-106-210S
10801-756EA 150.22 USD
10801-756
Anti-GPD1 Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-Glycerol-3-Phosphate Dehydrogenase recognizes the enzyme glycerol-3-phosphate dehydrogenase, a major component of lipid biosynthesis. Glyercol-3-phosphate dehydrogenase catalyzes the reduction of dihydroxyacetone phosphate (DHAP) to glycerol-3-phosphate. In addition, it assists in maintaining the redox potential across the inner mitochondrial membrane in glycloysis. Glycerol-3-phosphate dehydrogenase can be found in the cytosol and the inner mitochondrial membrane. Anti-Glycerol-3-Phosphate Dehydrogenase is ideal for investigators interested in metabolism, cancer, and cardiovascular diseases.

Recommended Dilutions: ELISA: 1:1,000 - 1:5,000; Western Blot: 1:500 - 1:2,500; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: GPD1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR