Anti-GPD1 Goat Polyclonal Antibody (BAC)

Supplier: Rockland Immunochemical
200-106-210-0100
10801-754EA 505.47 USD
10801-754
Anti-GPD1 Goat Polyclonal Antibody (BAC)
Antibodies
Anti-Glycerol-3-Phosphate Dehydrogenase recognizes the enzyme glycerol-3-phosphate dehydrogenase, a major component of lipid biosynthesis. Glyercol-3-phosphate dehydrogenase catalyzes the reduction of dihydroxyacetone phosphate (DHAP) to glycerol-3-phosphate. In addition, it assists in maintaining the redox potential across the inner mitochondrial membrane in glycloysis. Glycerol-3-phosphate dehydrogenase can be found in the cytosol and the inner mitochondrial membrane.

Anti-Glycerol-3-Phosphate Dehydrogenase is ideal for investigators interested in metabolism, cancer, and cardiovascular diseases.

Anti-Glycerol-3-Phosphate Dehydrogenase has been assayed against 1.0 ug of Glycerol-3-Phosphate-Dehydrogenase in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature.  A working dilution of 1:1,000 to 1:5,000 of the reconstitution concentration is suggested for this product.

Type: Primary
Antigen: GPD1
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR