Anti-GLPK Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
200-106-211S
10801-752EA 151.49 USD
10801-752
Anti-GLPK Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-Glycerol Kinase antibody recognizes the phosphotransferase enzyme glycerol kinase. Glycerol kinase catalyzes the formation of glycerol phosphate by transferring a phosphate from ATP to glycerol. Dihydroxyactone and L-glycerladehyde can also act as acceptors in place of glycerol. Anti-Glycerol Kinase is ideal for investigators interested in Cancer, Metabolism, and Kinase/Phosphatase enzymology.

Recommended Dilutions: ELISA: 1:1,000 - 1:5,000; Western Blot: 1:500 - 1:2,500; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: glpK
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Glycerol Kinase [Cellulomonas Species]
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR