Anti-GCG Mouse Monoclonal Antibody [clone: IMD-7]

Supplier: Rockland Immunochemical
200-301-P38
10801-744EA 571.49 USD
10801-744
Anti-GCG Mouse Monoclonal Antibody [clone: IMD-7]
Antibodies
Glucagon is a member of a multigene family that includes secretin. Glucagon is a 29-amino acid pancreatic hormone that counteracts the glucose-lowering action of insulin by stimulating glycogenolysis and gluconeogenesis. the human glucagon gene is approximately 9.4 kb long and contains 6 exons and 5 introns, and assigned to 2q36-2q37. This antibody is suitable for researchers interested in cancer research, metabolic diseases, transcription factors and nuclear receptors, neuroscience research, SKT signaling, GCPR signaling, and calcium signaling research.

Recommended Dilutions: Immunohistochemsitry: 1:100-1:500 ; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: GCG
Clonality: Monoclonal
Clone: IMD-7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR