Human Recombinant Glial Derived NTF
Supplier: Rockland Immunochemical
009-001-V10-0010 009-001-V10-0100
10801-702EA 8502 USD
10801-702 10801-708 10801-714 10801-720
Human Recombinant Glial Derived NTF
Proteins and Peptides
Recombinant Human Glial Derived NTF control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Glial Cell Line-Derived Neurotrophic Factor (GDNF) is a neurotrophic factor that is closely related to other neurotrophic factors, such as Neurturin, Persephin, and Artemin, by a common structural feature called the cysteine-knot. GDNF signals through a multicomponent system of receptors that includes RET and GFRα1-4, to promote dopamine uptake, survival and differentiation of neurons. Recombinant human GDNF is a non-glycosylated homodimer, containing two 135 amino acid chains, with a total molecular weight of 30.4 kDa.
Glial Derived Neurotrophic Factor Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Glial Derived Neurotrophic Factor in immunological assays.