Anti-BACE2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-G82
10801-680EA 635.22 USD
10801-680
Anti-BACE2 Rabbit Polyclonal Antibody
Antibodies
BACE2 antibody detects human amyloid-beta (Abeta) plaque. Accumulation of the amyloid-beta (Abeta) plaque in the cerebral cortex is a critical event in the pathogenesis of Alzheimer’s disease. Abeta peptide is generated by proteolytic cleavage of the beta-amyloid protein precursor (APP) at beta- and gamma-sites by proteases. The long-sought beta-secretase was recently identified by several groups independently and designated beta-site APP cleaving enzyme (BACE) and aspartyl protease 2 (Asp2). A BACE homolog was recently cloned and designated BACE2, Asp1, DRAP (for Down region aspartic protease), and memapsin 1. BACE2 also cleaves APP at beta-site and at a different site within Abeta. BACE2 locates on chromosome 21q22.3, the so-called ‘Down critical region’, suggesting that BACE2 and Abeta may also contribute to the pathogenesis of Down syndrome. Anti-BACE2 antibodies are ideal for investigators involved in Neuroscience and DNA Damage and Repair.

  • Recommended Dilutions: Western Blot: 0.5-1 ug/mL; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: BACE2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR