Anti-H5N1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-Z13
10801-670EA 635.22 USD
10801-670
Anti-H5N1 Rabbit Polyclonal Antibody
Antibodies
Influenza A virus is a major public health threat, killing more than 30, 000 people per year in the USA. Novel influenza virus strains caused by genetic drift and viral recombination emerge periodically to which humans have little or no immunity, resulting in devastating pandemics. Influenza A can exist in a variety of animals; however, it is in birds that all subtypes, including the so-called "avian flu" or H5N1, can be found. These subtypes are classified based on the combination of the virus coat glycoproteins hemagglutinin (HA) and neuraminidase (NA) subtypes. One of the less studied proteins encoded by the influenza virus is the nonstructural protein (NS) 2. NS2 binds to the influenza matrix protein M1 that is bound to the ribonucleoprotein (RNP) complex and mediates the contact between the M1/RNP complex and the cellular exportin CRM, but does not interact with nucleosomes.

Recommended Dilutions: ELISA: User Optimized ; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: H5N1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype:
Reactivity: Virus, H5N1 Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR