Human Recombinant Fractalkine
Supplier: Rockland Immunochemical
009-001-H49-0100 009-001-H49-0020 009-001-H49-0005
10801-478EA 4839 USD
10801-478 10801-480 10801-482 10801-484
Human Recombinant Fractalkine
Proteins and Peptides
rHuman Fractalkine (CX3CL1) control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Fractalkine, also known as CX3CL1, is an atypical chemokine that was the first of a fourth chemokine motif (CX3C). It is thought to function as a T cell and monocyte chemotractant and is produced by non-haemopoietic cells. Fractalkine is made in a soluble and membrane bound form in activated endothelial cells which is thought to promote adhesion of leukocytes. Recombinant human Fractalkine is a non-glycosylated protein, containing 76 amino acids, with a molecular weight of 8.6 kDa.
Fractalkine Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Fractalkine in immunological assays.