Anti-FOLR1 Goat Polyclonal Antibody (BAC)

Supplier: Rockland Immunochemical
200-106-148-0100
10801-444EA 531.79 USD
10801-444
Anti-FOLR1 Goat Polyclonal Antibody (BAC)
Antibodies
Anti-Folate Binding Protein antibody belongs to the folate receptor family and binds to folate and reduced folic acid derivatives. It further regulates delivery of 5-methyltetrahydrofolate to the cell interior. Folate expresses solely in epithelial tissues, and expression is increased in malignant tissues. Anti-Folate Binding Protein antibody is ideal for researchers investigating Tags & Cell Markers and Cancer research.

Anti-Folate Binding Protein has been assayed against 1.0 ug of Folate Binding Protein in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature.  A working dilution of 1:4,000 to 1:20,000 of the reconstitution concentration is suggested for this product.

Type: Primary
Antigen: FOLR1
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR