Anti-FMR1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-X07
10801-436EA 656.12 USD
10801-436
Anti-FMR1 Rabbit Polyclonal Antibody
Antibodies
Fragile X Mental Retardation Protein (FMRP) is an RNA-binding protein that plays an essential role in cognitive brain function. Mutations in the FMR1 gene, which codes for FMRP, can result in fragile X syndrome, autism, as well as other cognitive deficits. Phosphorylation of the highly conserved Ser499 has been shown to trigger hierarchical phosphorylation of nearby serines and may play a role in suppressing target mRNA translation. FMRP pS499 Antibody is ideal for researchers interested in neuroscience research.

Recommended Dilutions: ELISA: 1:10,000; Western Blot: 1:1000;

Type: Primary
Antigen: FMR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype:
Reactivity: Rat, Bovine, Canine, Chicken, Human, Mouse, Non-Human Primate, Sheep, Xenopus, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR