Mouse FGF basic
Supplier: Rockland Immunochemical
010-001-U80-0010 010-001-U80-0050
10801-368EA 2062 USD
10801-368 10801-370 10801-372
Mouse FGF basic
Proteins and Peptides
Recombinant Mouse FGF basic control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Fibroblast Growth Factors, FGFs, are a 22 member family of proteins known to be involved in angiogenesis, wound healing and embryonic development. As a family, they bind to heparin and signal through four receptor tyrosine kinases called, FGFR1, 2, 3 and 4. Although the mechanism remains unclear, FGF-basic, or FGF-1, is a critical component in keeping human embryonic stem cells undifferentiated in cell culture systems. Recombinant mouse FGF basic is a non-glycosylated protein, containing 145 amino acids, with a molecular weight of 16.3 kDa.
Fibroblast Growth Factor basic Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Fibroblast Growth Factor basic in immunological assays. Buffer Formulation: 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5.