Anti-Host-Cell-Proteins Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
200-401-M61S
10801-216EA 151.49 USD
10801-216
Anti-Host-Cell-Proteins Rabbit Polyclonal Antibody
Antibodies
Chemiluminescent Western Blot Kit: Anti-Host Cell Proteins Antibody recognizes high and low molecular weight protein fractions from E.coli. This antibody has been validated by 2D electrophoresis western blot to assure highest possible coverage of E.coli HCP. Host Cell Proteins are process-related impurities derived from a host cell expression system that may be present in trace amounts in a final drug substance. HCP Antibodies are used to detect impurities in biologically-based therapeutics. Detecting impurities often unseen by 1-D electrophoresis, a 2D electrophoresis western blot analysis delivers more complete insight into the immunocoverage of protein components of the host cell lysate.

Recommended Dilutions: ELISA: 1:1000-1:2000; Western Blot: 1:500-1:1000; contains 0.01% (w/v) Sodium Azide

Type: Kit
Antigen: Host-Cell-Proteins
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: E. coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR