Anti-CD55 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-AS3
10801-086EA 635.22 USD
10801-086
Anti-CD55 Rabbit Polyclonal Antibody
Antibodies
The decay-accelerating factor (DAF), also known as CD55, is an integral membrane glycoprotein that is involved in the regulation of the complement cascade (1). DAF limits excessive complement activation by accelerating their decay following DAF binding, thereby disrupting the cascade and preventing damage to host cells (2). Antigens present on the DAF glycoprotein constitute the Cromer blood group system (CROM) (3).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CD55
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR