Anti-CK18 Mouse Monoclonal Antibody [clone: CK-18]

Supplier: Rockland Immunochemical
200-301-P33
10801-076EA 571.49 USD
10801-076
Anti-CK18 Mouse Monoclonal Antibody [clone: CK-18]
Antibodies
Intermediate filaments (IFs) are a structurally related family of cellular proteins that appear to be intimately involved with the cytoskeleton. Human keratin 18(KRT18) and the homologous mouse Endo B are type I IF protein subunits whose expression is restricted in adults to a variety of simple epithelial tissues. The KRT18 gene is 3,791 bp long and the keratin 18 protein is coded for by 7 exons. The K18 gene is 3791 bp in length and the K18 protein is coded for by seven exons. By Southern blotting using the genomic DNA PCR product, the gene for keratin 18 is assigned to chromosome 12. Mutation of human keratin 18 in association with cryptogenic cirrhosis. This antibody is suitable for researchers interested in cancer research, cardiovascular diseases, and cytoskeletal signaling research.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 1:100-1:500; Western Blot: 0.5-1µg/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: CK18
Clonality: Monoclonal
Clone: CK-18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR