Human Recombinant CCNE1 (from Baculovirus (Sf9 Insect cells))
Supplier: Rockland Immunochemical
009-001-R79S
10800-970EA 714 USD
10800-970 10801-064
Human Recombinant CCNE1 (from Baculovirus (Sf9 Insect cells))
Proteins and Peptides
Cyclin E1 belongs to the highly conserved cyclin family, whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle that functions as regulators of CDK kinases. Cyclin E1 activity is required for centrosome duplication during S phase and this mechanism could coordinate centrosome reproduction with cycles of DNA synthesis and mitosis (1). The downregulation of cyclin E-CDK2 kinase activity following the G1/S-phase transition that is necessary for the maintenance of karyotypic stability. Cyclin E also has a modular centrosomal-targeting domain which is essential for promoting S phase entry in a Cdk2-independent manner (2). Cyclin E1 Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Cancer, and Cell Cycle research.