Anti-CXXC5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-AR7
10801-042EA 635.22 USD
10801-042
Anti-CXXC5 Rabbit Polyclonal Antibody
Antibodies
CXXC5 is zinc finger protein closely related to the DVL1-binding protein CXXC4 (also known as Idax), and like CXXC4 is thought to modulate the Wnt signaling pathway during development. CXXC5 is upregulated in neural stem cells upon BMP4 stimulation, leading to decreased levels of the Wnt-signaling target AXIN2 and facilitated the response of the stem cells to Wnt3a. Recent reports suggest CXXC5 transcription is activated by the Wilms tumor 1 (WT1) transcription factor, and together are involved in the WNT/beta-catenin pathway and play a role in embryonic kidney development.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 2.5 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CXXC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR