Anti-CRIM1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-AN3
10800-906EA 635.22 USD
10800-906
Anti-CRIM1 Rabbit Polyclonal Antibody
Antibodies
CRIM1 (cysteine-rich motor neuron 1), a glycosylated type I transmembrane protein, plays a role in tissue development i.e. capillary formation and maintenance during angiogenesis. It contains an N-terminal IGF-binding protein-like motif and six von Willebrand-like cysteine-rich repeats (CRRs) in its extracellular domain. CRIM1 interacts with BMP4 and BMP7 via the CRRs and functions as an antagonist. CRIM1 is developmentally expressed in a number of tissues including the pancreas, kidney, placenta, brain and blood vessels. CRIM1 may participate in CNS and placental development by interacting with growth factors involved in motor neuron differentiation and survival.

Recommended Dilutions: ELISA: 1:10,000; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CRIM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR