Anti-CCDC106 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-AF0
10800-626EA 635.22 USD
10800-626
Anti-CCDC106 Rabbit Polyclonal Antibody
Antibodies
The coiled-coil domain is a common protein motif that is often involved in protein oligomerization and is found in proteins such as transcription factors and intermediate filaments. CCDC106 was initially identified as a p53-interacting protein by yeast two-hybrid screening. Other experiments demonstrated that CCDC106 co-localizes and interacts with p53 in the nucleus, inhibiting the transcriptional activity of p53 and stimulating p53 protein degradation, indicating that at least one of the functions of CCDC106 is acting as a negative regulator of p53.

Recommended Dilutions: ELISA: 1:5000-1:10000; Immunohistochemsitry: 5 µg/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 0.5-1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CCDC106
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR