Bovine Serum Albumin (BSA)
Supplier: Rockland Immunochemical
BSA-5KG
10800-424EA 5775.52 USD
10800-424
Bovine Serum Albumin (BSA)
Proteins and Peptides
Lyophilized Bovine Serum Albumin - Fraction V, pH 7.0.
  • use in protease sensitive assays: RIA, EIA and nucleic acid hybridization
  • use as a stabilizing agent for proteins and enzymes
  • dilute solutions of antibody; use as a blocking agent to reduce non-specific binding
  • Purity (%): 100% by Agarose Zone electrophoresis
  • Protein (%): 100% by nitrogen analysis.
  • IgG, Protease, Heavy Metal Free.
Bovine Serum Albumin (BSA) is used for various biochemical applications including ELISA (Enzyme-Linked Immunosorbent Assay), high content screenining assays, western blotting, and immunohistochemistry. BSA as a blocking reagent is particularly useful with casein-sensitive antibodies, such as phospho-specific antibodies. Also used as a nutrient in cell and microbial culture. In restriction digests, BSA is used to stabilize some enzymes during digestion of DNA and to prevent adhesion of the enzyme to reaction tubes and other vessels. Bovine Serum Albumin can also be used to determine the quantity of other proteins, by comparing an unknown quantity of protein to known amounts of BSA.
This product is suitable for: use in protease sensitive assays such as RIA, EIA and nucleic acid hybridization, use as a stabilizing agent for proteins and enzymes including dilute solutions of antibody, use as a blocking agent to reduce non-specific binding.