Blocking Buffer Sampler Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
K505
10800-390EA 394.93 USD
10800-390
Blocking Buffer Sampler Kit, Rockland Immunochemicals
Buffers Electrophoresis Buffers Blocking Buffers
Blocking Buffer Sampler Kit for ELISA, immunoassays, and western blot
  • B304 - Normal Goat Serum
  • BSA-10 - Bovine Serum Albumin (BSA)
  • MB-070 - Blocking Buffer for Fluorescent Western Blotting
  • MB-064 - ELISA Microwell Blocking Buffer with Stabilizer
  • MB-067 - 10X TBS Fish Gel Concentrate (Azide and Mercury free)
Rockland lyophilized NORMAL GOAT SERUM (NGS) is recommended for use in immunoassays as a source of non-specific serum protein or on tissue for immunohistochemical applications
BOVINE SERUM ALBUMIN is suitable dilute solutions of antibody, use as a blocking agent to reduce non-specific binding
2X Blocking Buffer for Fluorescent Western Blotting is optimized for Fluorescent Westen blotting and other immunoassays
ELISA Microwell Blocking Buffer with Stabilizer (Azide and Mercury Free) for ELISA
10X TBS Fish Gel Concentrate (Azide and Mercury free) for western blot or ELISA
Application Note: Blocking Buffer Sampler Kit contains: NORMAL GOAT SERUM (NGS), BOVINE SERUM ALBUMIN - Fraction V (Immunoglobulin and Protease Free), Blocking Buffer (2X) for Fluorescent Western Blotting, ELISA Microwell Blocking Buffer with Stabilizer (Azide and Mercury Free) - Designed for ELISA assays, 10X TBS Fish Gel Concentrate (Azide and Mercury free) is a blocking agent for immunoassays and western blot.