Anti-Bilirubin Oxidase Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical
200-106-212S
10800-370EA 151.49 USD
10800-370
Anti-Bilirubin Oxidase Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-Bilirubin Oxidase antibody is an enzyme that increases the oxidation rate of bilirubin to biliverdin and other substrates. It is ideal for investigators interested in Metabolism or Signal Transduction research.

Recommended Dilutions: ELISA: 1:1,000 - 1:5,000; Immunoprecipitation: 1:100; Western Blot: 1:500 - 1:2,500; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: Bilirubin Oxidase
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Myrothecium Verrucaria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR