Anti-HA Goat Polyclonal Antibody

Supplier: Rockland Immunochemical
600-101-Z08
10800-264EA 635.22 USD
10800-264
Anti-HA Goat Polyclonal Antibody
Antibodies
Influenza A virus is a major public health threat, killing more than 30, 000 people per year in the USA. Novel influenza virus strains caused by genetic drift and viral recombination emerge periodically to which humans have little or no immunity, resulting in devastating pandemics. Influenza A can exist in a variety of animals; however it is in birds that all subtypes can be found. These subtypes are classified based on the combination of the virus coat glycoproteins hemagglutinin (HA) and neuraminidase (NA) subtypes. During 1997, an H5N1 avian influenza virus was determined to be the cause of death in 6 of 18 infected patients in Hong Kong. There was some evidence of human to human spread of this virus, but it is thought that the transmission efficiency was fairly low. HA interacts with cell surface proteins containing oligosaccharides with terminal sialyl residues. Virus isolated from a human infected with the H5N1 strain in 1997 could bind to oligosaccharides from human as well as avian sources, indicating its species-jumping ability.

Recommended Dilutions: ELISA: 1:10,000; Western Blot: 1 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: HA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Goat
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR