Human Recombinant Artemin
Supplier: Rockland Immunochemical
009-001-U64-0100 009-001-U64-0020 009-001-U64-0005
10800-088EA 4839 USD
10800-088 10800-090 10800-092 10800-094
Human Recombinant Artemin
Proteins and Peptides
Recombinant Human Artemin control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Artemin is a novel member of the glial cell line-derived neurotrophic factor (GNDF) ligand family. Current evidence suggests that Artemin signals through the receptor complex, GFRα3-RET, to influence neuron survival in vitro and in vivo. Recombinant human Artemin is a non-glycosylated, disulfide-linked homodimer, containing two 113 amino acid chains, with a total molecular weight of 24.2 kDa.
Recombinant human Artemin is suitable as a control for polyclonal or monoclonal anti-Artemin in immunological assays.