Anti-AMBRA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-Y09
10799-990EA 635.22 USD
10799-990
Anti-AMBRA1 Rabbit Polyclonal Antibody
Antibodies
Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. Beclin-1, a principal regulator of autophagosome formation, is in turn regulated by Ambra1. Ambra1 associates with Beclin-1 through a region near its center as determined by yeast two-hybrid assay. Null mutations in this gene in mice resulted in embryonic lethality with severe neural tube defects associated with autophagy impairment, accumulation of ubiquitinated proteins, unbalanced cell proliferation and excessive apoptotic death. Furthermore, down-regulation of Ambra1 in cultured cells though RNA interference decreased the level of rapamycin- and nutrient starvation-induced autophagy. Multiple isoforms of Ambra1 are known to exist.

Recommended Dilutions: ELISA: 1:20,000; Immunohistochemsitry: 5 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 2 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: AMBRA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR