Anti-5-mC Mouse Monoclonal Antibody [clone: 33D3]

Supplier: Rockland Immunochemical
200-301-V02
10799-862EA 871.34 USD
10799-862
Anti-5-mC Mouse Monoclonal Antibody [clone: 33D3]
Antibodies
The 5-methylcytosine antibody (clone 33D3) is the most published and widely used antibody for DNA methylation analysis. Anti-5mC is ideal for research in Epigenetics due to its applicability to DNA methylation analysis.

Recommended Dilutions: ELISA: 1:3,000; ChIP: 1 ?g/ChIP; Western Blot: 1:1,000; contains 0.05% (w/v) Sodium Azide

Type: Primary
Antigen: 5-mC
Clonality: Monoclonal
Clone: 33D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Vertebrates, Plants
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR