Trajan, PEEK Fingertight Fittings, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
222017
10799-764PK 95.46 USD
10799-764
Trajan, PEEK Fingertight Fittings, Trajan Scientific and Medical
Fittings and Connectors
PEEK Fingertight fittings offer absolute zero to low dead volume connections and are ideal for bioanalytical applications.
  • Unique design provides Absolute Zero or low dead volume connections
  • Biocompatible material