Chemical/Abrasion-Resistant Cleanroom Labels, CleanMark®
Supplier: Lgi
L65287
10799-278CS 1213.97 USD
10799-278
Chemical/Abrasion-Resistant Cleanroom Labels, CleanMark®
Labels
Chemical/abrasion cleanroom labels are recommended for use in critical environments.
Chemical/abrasion cleanroom labels are designed for critical environments that need protection from chemicals and/or abrasion. Lgi offers a variety of materials, adhesives, and over laminates that can be used in the harshest environments where resistance to moisture, chemicals, solvents, and cleaning agents are in use.
CleanMark® labels can be custom constructed to fit any size, and can be used with either direct thermal or thermal transfer printers. These labels can withstand extreme hot or cold temperatures, harsh chemical solvents, and physical abrasion. They can indicate sterilization processes such as autoclaving, may be pre-sterilized, can firmly adhere to or easily peel from challenging surfaces, and will not contaminate clean or sterile environments with particulates, outgassing, or microbes. Please call your VWR Sales Representative for additional information.
Certifications: RoHs compliant.