VWR® Balance Interface Cable
Supplier: VWR International
10799-176
10799-176EA 151.52 USD
10799-176
VWR® Balance Interface Cable
Cables
Allows for connection between a VWR® balance and computer.
The interface cable allows for a connection between a VWR balance and a computer. The cable features a DB9-F connection (balance) to USB-A (computer). Additional software may be required to record data into computer. Compatible VWR® balances include: A-series, T-series, B2-series, B2T-series, P2-series, E-series, VWR® moisture analyzers.