VWR® Compact Micro Centrifuges
Supplier: VWR International
10799-122 10799-170 10799-174 76019-134 76019-136 76019-138 76019-204 76019-206 76019-208
10799-122EA 5236.22 USD
10799-122 10799-170 10799-174 76019-134 76019-136 76019-138 76019-204 76019-206 76019-208
VWR® Compact Micro Centrifuges
Centrifuges Microcentrifuges
These compact microcentrifuges are ideal for DNA, RNA, and temperature-sensitive samples.
  • Refrigerated Model with Pre-Cool Function Available
  • Robust and Steady Temperature Control
  • High Performance; Ideal for Sample Preparation
  • Short Run-up and Run-down Times
  • Pulse Button for Short Centrifugation Runs
Safety locking lids are equipped for biocontainment.
They are autoclavable and phenol-resistant. Operation noise made by these units is less than 58 dB, which is softer than a normal conversation.
Ordering information: Units come with a five-year manufacturer's warranty that extends the standard manufacturer's two-year warranty for an additional three years. The three-year extension is a full powertrain manufacturer's warranty on the centrifuge only. A comprehensive range of rotor options is available separately.