Steam Dot Blu™ Process Indicator, Propper Manufacturing
Supplier: Propper
36400200
10798-796PK 142.26 USD
10798-796
Steam Dot Blu™ Process Indicator, Propper Manufacturing
Sterilization Indicators
Steam-Dot™ Indicators provide a fast-positive visual check for processed packages sterilized in steam.
  • Self-adhesive steam process indicator
  • Permanent white to black color change on exposure to steam
  • Decipher between processed and unprocessed packages
The dot has a 1.27 cm (½") diameter, which is ideal for placement on sterilization container locks. Propper manufactures the Steam Dot Blu to be lead-free and heavy metal-free.
Certifications: AAMI ST60 and ISO 11140-1, Type 1