DualClean™ Softwall Strip Cleaner, Contec
Supplier: Contec
2778 2746S DCQT0001 DCMD0001 DCQT0002 P191061E
10797-032EA 429.91 USD
10797-032 10797-034 10797-036 10797-038 76121-376 77282-228
DualClean™ Softwall Strip Cleaner, Contec
Mops
The DualClean™ Softwall Strip Cleaner is ideal for cleaning strip curtains and barriers in clean workspace enclosures and ISO Class 5 isolated cleanroom work cells.
  • Stainless steel frame securely holds wiping covers for uniform strip contact
  • Cleans both sides of strip curtain, up to 12" wide in one pass
  • Tension on strip is easily controlled
  • Prevents cleaning covers from twisting and binding
  • Can be sterilized by most common methods, making it ideal for critical environments
  • Traps and removes contaminants
  • Extremely clean and durable
  • Compatible with most
More Product Information