Anti-DEFB132 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX53990
10795-268EA 623.1 USD
10795-268
Anti-DEFB132 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to DEFB132

Type: Primary
Antigen: DEFB132
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR