Anti-TBL1XR1 Mouse Monoclonal Antibody [clone: 4F3-A8-D9]

Supplier: Genetex
GTX49189
10795-118EA 615.49 USD
10795-118
Anti-TBL1XR1 Mouse Monoclonal Antibody [clone: 4F3-A8-D9]
Antibodies
Mouse Monoclonal antibody [4F3-A8-D9] to TBLR1

Type: Primary
Antigen: TBL1XR1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR