Anti-wnt8b Rabbit Polyclonal Antibody

Supplier: Genetex
GTX54245
10795-074EA 708.44 USD
10795-074
Anti-wnt8b Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Wnt8b

Type: Primary
Antigen: wnt8b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Zebra Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR