Anti-ITGA1 Mouse Monoclonal Antibody [clone: TS2/7]

Supplier: Genetex
GTX15767
10794-380EA 628.33 USD
10794-380
Anti-ITGA1 Mouse Monoclonal Antibody [clone: TS2/7]
Antibodies
Mouse monoclonal antibody [TS2/7] to Integrin alpha 1

Type: Primary
Antigen: ITGA1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR