Anti-RAB32 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX130477
10793-538EA 589.27 USD
10793-538
Anti-RAB32 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to RAB32 (RAB32, member RAS oncogene family)

Type: Primary
Antigen: RAB32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR