Anti-STX17 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX130212
10793-526EA 589.27 USD
10793-526
Anti-STX17 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to STX17 (syntaxin 17)

Type: Primary
Antigen: STX17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR