Anti-KDR Rabbit Polyclonal Antibody

Supplier: Genetex
GTX129943
10793-496EA 589.27 USD
10793-496
Anti-KDR Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to VEGFR-2 (kinase insert domain receptor (a type III receptor tyrosine kinase))

Type: Primary
Antigen: KDR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR