Anti-PCBD1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX54554
10792-760EA 623.1 USD
10792-760
Anti-PCBD1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PCBD1

Type: Primary
Antigen: PCBD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR