Anti-PRKRA Rabbit Polyclonal Antibody

Supplier: Genetex
GTX54546
10792-752EA 623.1 USD
10792-752
Anti-PRKRA Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PACT

Type: Primary
Antigen: PRKRA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR