Anti-ITGB2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX54372
10792-728EA 623.1 USD
10792-728
Anti-ITGB2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CD18

Type: Primary
Antigen: ITGB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR